Learn More
Sino Biological Anti-EDA2R/TNFRSF27 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11845RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human XEDAR / EDA2R (rh XEDAR / EDA2R; Catalog#11845-H08H; NP_068555.1; Met1-Thr138). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4521