Learn More
Sino Biological Anti-Endothelin A Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104886T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Endothelin A Receptor/EDNRA, and purified by antigen affinity chromatography.
Catalog No. 50-241-0930