Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human EFCAB4A Synthetic Peptide

Catalog No. PEP1173 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1173 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1173 Supplier Invitrogen™ Supplier No. PEP1173
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 15 amino acids near the center of human EFCAB4A. PEP-1173 can be used as a blocking peptide with polyclonal antibody PA5-21059.

EFCAB4A, also known as Calcium release-activated calcium channel regulator 2B, is a novel Ca2+-binding EF-hand protein that is thought to play a key role in store-operated Ca2+ entry in T-cells by regulating CRAC channel activation, but the detailed function is still under investigation. It is likely to play a similar role as the related protein EFCAB4B, which acts as a cytoplasmic calcium-sensor that forms a complex with ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca2+ concentration.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 283229
Species Human
Purification Method purified
Accession Number Q8N4Y2
Common Name EFCAB4A
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 6330520A15Rik; BC026645; Ca2+ release-activated Ca2+ (CRAC) channel regulator 2B; calcium release activated channel regulator 2B; calcium release-activated calcium channel regulator 2B; Calcium release-activated channel regulator 2B; CRAC channel regulator 2B; CRAC regulator 2B; Cracr2a; CRACR2B; Efcab4; EFCAB4A; EF-hand calcium binding domain 4A; EF-hand calcium-binding domain-containing protein 4A; RGD1560911
Gene Symbol CRACR2B
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.