Learn More
Sino Biological Recombinant Anti-Ephrin-A1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10882R453100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Ephrin-A1 / EFNA1 (rh Ephrin-A1 / EFNA1; Catalog#10882-H08H; NP_004419.2; Met1-Ser182).
Catalog No. 50-241-0941