Learn More
Sino Biological Anti-Ephrin-A1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10882MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EphrinA1 (rh EphrinA1; Catalog#10882-H08H; NP_004419.2; Met 1-Ser 182). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0518