Learn More
Sino Biological Anti-Ephrin-A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10882RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EphrinA1 / EFNA1 (rh EphrinA1; Catalog#10882-H08H; NP_004419.2; Met 1-Ser182). EphrinA1 specific IgG was purified by human EphrinA1 affinity chromatography.
Catalog No. 50-240-8670