Learn More
Sino Biological Human EFNA4 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 5A0476M08H1
Encompass_Preferred
A DNA sequence encoding the human EFNA4 (NP_005218.1)(Met1-Gly171) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4353