Learn More
FUJIFILM Biosciences Human EG-VEGF, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50900459
Encompass_Preferred
Endocrine gland-derived vascular endothelial growth factor (EG-VEGF) is an angiogenic growth factor that is expressed in the ovaries, testis, adrenal, and placental tissues.   EG-VEGF has mitogenic, chemoattractive, and antiapoptotic functional roles.  EG-VEGF signaling is mediated through binding the G protein-coupled receptors prokineticin receptor 1 (PKR1) and prokineticin receptor 2 (PKR2).  Polycystic ovaries display strong EG-VEGF expression that is associated with increased angiogenesis and cyst formation, which could lead to the formation of polycystic ovary syndrome and infertility. 
Catalog No. 50-900-459 Supplier FUJIFILM Biosciences Supplier No. 10007100UG
Add to Cart
Edge
Add to Cart
Endocrine gland-derived vascular endothelial growth factor (EG-VEGF) is an angiogenic growth factor that is expressed in the ovaries, testis, adrenal, and placental tissues.   EG-VEGF has mitogenic, chemoattractive, and antiapoptotic functional roles.  EG-VEGF signaling is mediated through binding the G protein-coupled receptors prokineticin receptor 1 (PKR1) and prokineticin receptor 2 (PKR2).  Polycystic ovaries display strong EG-VEGF expression that is associated with increased angiogenesis and cyst formation, which could lead to the formation of polycystic ovary syndrome and infertility. 