Learn More
Sino Biological Human EGF MAb 500ug (Sino Biological)
Supplier:  Sino Biological 10605R001500
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EGF / Epidermal Growth Factor (rh EGF / Epidermal Growth Factor; Catalog#10605-HNAE; NP_001954.2; Asn971-Arg1023) and was produced using recombinant antibody technology.
Catalog No. 50-237-0797