Learn More
Sino Biological Anti-EGF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10605RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EGF / Epidermal Growth Factor (rh EGF; Catalog#10605-HNAE; NP_001954.2; Asn971-Arg1023). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1103