Learn More
ACROBiosystems PE-Labeled Human EGF R Protein, His TagStar Staining
Catalog No. 502331283
Encompass_Preferred
PE-Labeled Human EGF R Protein, His Tag (EGR-HP2H7) is produced via conjugation of PE to Human EGF R Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGF R Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).
Catalog No. 50-233-1283 Supplier ACROBiosystems Supplier No. EGRHP2H725TESTS
PE-Labeled Human EGF R Protein, His Tag (EGR-HP2H7) is produced via conjugation of PE to Human EGF R Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGF R Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).