Learn More
ACROBiosystems Human EGFR-coupled Magnetic Beads
Catalog No. 502331423
Encompass_Preferred
Human EGFR protein is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645(Accession # P00533-1).
Catalog No. 50-233-1423 Supplier ACROBiosystems Supplier No. MBEK0122MG
Human EGFR protein is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645(Accession # P00533-1).