Learn More
ACROBiosystems ACROBiosystems Human EGF R Protein, Llama IgG2b Fc Tag
Catalog No. 502106919
Encompass_Preferred
Recombinant Protein;100UG;Human EGF R Protein, Llama IgG2b Fc Tag (EGR-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 96.4 kDa.
Catalog No. 50-210-6919 Supplier ACROBiosystems Supplier No. EGRH5259100UG
Recombinant Protein;100UG;Human EGF R Protein, Llama IgG2b Fc Tag (EGR-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 96.4 kDa.