Learn More
Proteintech Group Inc Human EGFR ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0014296T
Encompass_Preferred
The epidermal growth factor receptor (EGFR) belongs to the ErbB family of receptor tyrosine kinases (RTKs) and exerts critical functions in epithelial cell physiology. EGFR is a receptor of tyrosine kinase (RTK). It is consisted of a C-terminus intracellular region that possesses the kinase activity, and an N-terminus extracellular ligand-binding site, a hydrophobic transmembrane domain. EGFR is frequently mutated and/or overexpressed in different types of human cancers and is the target of multiple cancer therapies currently adopted in the clinical practice. Mutations in this gene are associated with lung cancer.
Catalog No. 50-197-4100