Learn More
Sino Biological Human EGFR ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370710
Encompass_Preferred
Each vial contains 140 ng of recombinant Human EGFR/HER1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 3000 pg/mL is recommended.
Catalog No. 50-237-0710 Supplier Sino Biological Supplier No. SEK1000115
Each vial contains 140 ng of recombinant Human EGFR/HER1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 3000 pg/mL is recommended.