Learn More
Sino Biological Recombinant Human EGFR Protein (Isoform vIII, His Tag) 1
Supplier:  Sino Biological 29662H08B1
Encompass_Preferred
A DNA sequence encoding the human EGFRvIII (NP_001333870.1) (Met1-Lys375) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6650