Learn More
Sino Biological Recombinant Anti-EGFR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10001RE11100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EGFR / HER1 (rh EGFR / HER1; Catalog#10001-H08H; NP_005219; Met1-Ser645).
Catalog No. 50-241-2383