Learn More
Sino Biological Phospho-EGF Receptor (Tyr845) Antibody, Rabbit MAb
Supplier:  Sino Biological 110828R000720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Tyr845 of the Human Phospho-EGF Receptor.
Catalog No. 50-255-4855