Learn More
Sino Biological Anti-EGFR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10001RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EGFR / ErbB / HER1 Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5551