Learn More
Sino Biological Recombinant Human EGFR Protein (ECD, Isoform vIII, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 29662H27BB100
Encompass_Preferred
A DNA sequence encoding the human EGFRvIII (NP_001333870.1) (Met1-Ser378) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3828