Learn More
ACROBiosystems APC-Labeled Human EGFRvIII, His Tag (EGI-HA2H8) is produced via conjugation of APC to Human EGFRvIII, His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human EGFRvIII, His Tag is
Supplier:  ACROBiosystems EGIHA2H8200TESTS
Encompass_Preferred
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.The type III EGF deletion-mutant receptor (EGFRvIII) is the most common mutation and was first identified in primary human glioblastoma tumors; EGFR gene amplification is correlated with the structural rearrangement of the gene. The EGFRvIII gene has an in-frame deletion of 801 base pairs, corresponding to exons 2–7 in the mRNA, resulting in the deletion of amino acids 30-297 in the extracellular domain and the generation of a glycine at the fusion point
Catalog No. 50-225-4046