Learn More
ACROBiosystems PE-Labeled Human EGFRvIII Protein, His TagStar Staining
Catalog No. 502331379
Encompass_Preferred
PE-Labeled Human EGFRvIII, His Tag (EGI-HP2H6) is produced via conjugation of PE to Human EGFRvIII, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGFRvIII, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).
Catalog No. 50-233-1379 Supplier ACROBiosystems Supplier No. EGIHP2H625TESTS
PE-Labeled Human EGFRvIII, His Tag (EGI-HP2H6) is produced via conjugation of PE to Human EGFRvIII, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGFRvIII, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).