Learn More
ACROBiosystems APC-Labeled Human EGFRvIII Protein, His TagStar Staining
Supplier:  ACROBiosystems EGIHA2H525TESTS
Encompass_Preferred
APC-Labeled Human EGFRvIII, His Tag (EGI-HA2H5) is produced via conjugation of APC to Human EGFRvIII, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGFRvIII, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).
Catalog No. 50-233-1362