Learn More
ACROBiosystems Alexa Fluor 488-Labeled Human EGFRvIII Protein, His TagStar Staining
Catalog No. 502331580
Encompass_Preferred
Alexa Fluor 488-Labeled Human EGFRvIII, His Tag (EGI-HA2H7) is produced via conjugation of AF488 to Human EGFRvIII, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGFRvIII, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).
Catalog No. 50-233-1580 Supplier ACROBiosystems Supplier No. EGIHA2H725UG
Alexa Fluor 488-Labeled Human EGFRvIII, His Tag (EGI-HA2H7) is produced via conjugation of AF488 to Human EGFRvIII, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGFRvIII, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1).