Learn More
Raybiotech Inc Human EGR1 ELISA Kit
Supplier:  Raybiotech Inc ELHEGR11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 51 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human EGR1.
Catalog No. 50-194-9213