Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EIF2AK4 (NP_Q9P2K8.2, 1 a.a. - 1649 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells

Catalog No. 89943001 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-001 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-943-001 Supplier Abnova™ Supplier No. P5554
Only null left
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

EIF2AK4 belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2S1; MIM 603907) to downregulate protein synthesis in response to varied cellular stresses (Berlanga et al., 1999 [PubMed 10504407]).[supplied by OMIM]

Specifications

Accession Number NP_Q9P2K8.2
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 440275
Molecular Weight (g/mol) 215kDa
Name EIF2AK4 (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing Loading 1 ug protein in SDS-PAGE
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GCN2/KIAA1338
Common Name EIF2AK4
Gene Symbol EIF2AK4
Biological Activity The activity was determined by ELISA. The enzyme was incubated with GST-fused substrate protein and tRNA, and after stopping kinase reaction by EDTA, the reaction solution was transferred into glutathione-coated plate. Phosphorylation was detected by anti- phospho antibody and HRP-labeled anti-rabbit IgG. Substrate: EIF2S1. ATP: 100 uM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 90% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.