Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EIF3S12 Full-length ORF (NP_037366.1, 1 a.a. - 218 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7100077 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-046 10 μg
89-943-047 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-943-046 Supplier Abnova™ Supplier No. H00027335P01S
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The 700-kD eukaryotic translation initiation factor-3 (eIF3) is the largest eIF and contains at least 12 subunits, including EIF2S12. eIF3 plays an essential role in translation by binding directly to the 40S ribosomal subunit and promoting formation of the 40S preinitiation complex (Mayeur et al., 2003 [PubMed 14519125]).[supplied by OMIM]

Sequence: MAMFEQMRANVGKLLKGIDRYNPENLATLERYVETQAKENAYDLEANLAVLKLYQFNPAFFQTTVTAQILLKALTNLPHTDFTLCKCMIDQAHQEERPIRQILYLGDLLETCHFQAFWQALDENMDLLEGITGFEDSVRKFICHVVGITYQHIDRWLQAEMLGDLSDSQLKVWMSKYGWSADESGQIFICSQEESIKPKNIVEKIDFDSVSSIMASSQ

Specifications

Accession Number NP_037366.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27335
Molecular Weight (g/mol) 51.5kDa
Name EIF3S12 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAMFEQMRANVGKLLKGIDRYNPENLATLERYVETQAKENAYDLEANLAVLKLYQFNPAFFQTTVTAQILLKALTNLPHTDFTLCKCMIDQAHQEERPIRQILYLGDLLETCHFQAFWQALDENMDLLEGITGFEDSVRKFICHVVGITYQHIDRWLQAEMLGDLSDSQLKVWMSKYGWSADESGQIFICSQEESIKPKNIVEKIDFDSVSSIMASSQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ARG134/EIF3-p28/EIF3S12/HSPC029/M9/MSTP001/PLAC-24/PLAC24/PRO1474/PTD001
Common Name EIF3K
Gene Symbol EIF3K
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.