Learn More
Sino Biological Anti-EIF4H Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107059T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human EIF4H, and purified by antigen affinity chromatography.
Catalog No. 50-241-1586