Learn More
Biomatik Corporation Human Elastase 3A (ELA3A) ELISA Kit
Supplier:  Biomatik Corporation EKU03847
Encompass_Preferred
Human Elastase 3A (ELA3A) ELISA Kit for detecting ELA3A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09093. Gene ID: 10136. Target Synonyms: CELA3A; ELA3; Chymotrypsin-Like Elastase Family Member 3A; Protease E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8999