Learn More
Biomatik Corporation Human Elastin Microfibril Interface Located Protein 2 (EMILIN2) CLIA Kit
Supplier:  Biomatik Corporation EKU10044
Encompass_Preferred
Human Elastin Microfibril Interface Located Protein 2 (EMILIN2) CLIA Kit for detecting EMILIN2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 8.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9BXX0. Gene ID: 84034. Target Synonyms: FOAP-10; Elastin Microfibril Interfacer 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8378