Learn More
Biomatik Corporation Human ELAV Like Protein 1 (ELAVL1) ELISA Kit
Supplier:  Biomatik Corporation EKU09277
Encompass_Preferred
Human ELAV Like Protein 1 (ELAVL1) ELISA Kit for detecting ELAVL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15717. Gene ID: 1994. Target Synonyms: ELAV1; HUR; Hua; MelG; Hu Antigen R; Embryonic Lethal,Abnormal Vision Like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7745