Learn More
Biomatik Corporation Human Electron Transfer Flavoprotein Alpha Polypeptide (ETFa) ELISA Kit
Supplier:  Biomatik Corporation EKU03860
Encompass_Preferred
Human Electron Transfer Flavoprotein Alpha Polypeptide (ETFa) ELISA Kit for detecting ETFa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13804. Gene ID: 2108. Target Synonyms: EMA; GA2; MADD; Glutaric Aciduria II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4692