Learn More
Hycult Biotech Inc Elastase, Human, ELISA kit, 2 x 96 det.””
SDP
Supplier:  Hycult Biotech Inc HK31902
Encompass
The human neutrophil elastase ELISA has been developed for the quantitative measurement of free and bound natural human neurophil elastase in cell culture medium and plasma.
Catalog No. NC0548151