Learn More
BIOHIPPO Human ELN Microsample
SDP
Catalog No. 5000314421
Encompass
Human ELN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELN in serum plasma tissue homogenates and other biological fluids It provides a detection range of 4 69-300 ng/mL with a sensitivity of 0 17 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ELN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ELN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ELN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14421 Supplier BIOHIPPO Supplier No. BHE15212169
Human ELN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELN in serum plasma tissue homogenates and other biological fluids It provides a detection range of 4 69-300 ng/mL with a sensitivity of 0 17 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ELN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ELN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ELN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color