Learn More
ACROBiosystems Human Elongin B & Elongin C & VHL Heterotrimer Protein, Strep II and His tagged, 500 μg
Catalog No. 502370261
Encompass_Preferred
Recombinant human Elongin B, Elongin C, and VHL co-expressed as a heterotrimeric complex with Strep II and His affinity tags. Supplied as 500 μg and verified by MALS and SPR for use in biochemical and biophysical assays.
  • Heterotrimeric complex of human Elongin B, Elongin C, and VHL.
  • Includes Strep II and His affinity tags for purification.
  • Verified by MALS and SPR for molecular mass and binding analyses.
  • Supplied as 500 μg for biochemical and biophysical experiments.
  • Suitable for protein-protein interaction and structural studies.
Catalog No. 50-237-0261 Supplier ACROBiosystems Supplier No. ELLH5595500UG
Recombinant human Elongin B, Elongin C, and VHL co-expressed as a heterotrimeric complex with Strep II and His affinity tags. Supplied as 500 μg and verified by MALS and SPR for use in biochemical and biophysical assays.
  • Heterotrimeric complex of human Elongin B, Elongin C, and VHL.
  • Includes Strep II and His affinity tags for purification.
  • Verified by MALS and SPR for molecular mass and binding analyses.
  • Supplied as 500 μg for biochemical and biophysical experiments.
  • Suitable for protein-protein interaction and structural studies.