Learn More
ACROBiosystems ACROBiosystems Human EMMPRIN / CD147 Protein, Fc Tag
Supplier:  ACROBiosystems CD7H5259100UG
Encompass_Preferred
Recombinant Protein;100UG;Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EMMPRIN, Fc Tag (Cat. No. CD7-H5259) at 1 μg/mL (100 μL/well) can bind Anti-EMMPRIN Antibody, Mouse IgG1 with a linear range of 0.2-4 ng/mL (QC tested).
Catalog No. 50-210-6570