Learn More
BIOHIPPO Human EMMPRIN Microsample
SDP
Catalog No. 5000315589
Encompass
Human EMMPRIN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EMMPRIN in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 13 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human EMMPRIN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human EMMPRIN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human EMMPRIN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15589 Supplier BIOHIPPO Supplier No. BHE15212572
Human EMMPRIN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EMMPRIN in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 13 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human EMMPRIN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human EMMPRIN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human EMMPRIN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color