Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human EN-RAGE/S100A12 ELISA Kit

Catalog No. EH170RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH170RB 96 Tests
EH170RBX5 5 x 96 Tests
EH170RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH170RB Supplier Invitrogen™ Supplier No. EH170RB
Add to Cart
Edge
Add to Cart

ELISA

Human EN-RAGE/S100A12 quantitates human EN-RAGE/S100A12 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human EN-RAGE/S100A12.

S100A12 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100A12 is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities. S100A12 is also a ligand for RAGE. Interaction with RAGE on mononuclear phagocytes, lymphocytes and endothelium triggers cellular activation. Nuclear factor kappa B, a key transcription factor for inflammatory events, is activated by S100A12/RAGE. S100A12 has also been shown to have anti-microbial properties.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80511
Assay Range 0.82-600 ng/mL
Assay Sensitivity 0.82 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CAAF1; CAGC; Calcitermin; calcium-binding protein in amniotic fluid 1; calgranulin C; calgranulin-C; CGRP; EN RAGE; ENRAGE; EN-RAGE; Extracellular newly identified RAGE-binding protein; migration inhibitory factor-related protein 6; MRP6; MRP-6; Neutrophil S100 protein; p6; Protein S100 A12; protein S100-A12; S100 calcium binding protein A12; S100 calcium-binding protein A12; S100 calcium-binding protein A12 (calgranulin C); S100A12CAAF1; CAGC; Calcitermin; calcium-binding protein in amniotic fluid 1; calgranulin C; calgranulin-C; CGRP; EN RAGE; ENRAGE; EN-RAGE; Extracellular newly identified RAGE-binding protein; migration inhibitory factor-related protein 6; MRP6; MRP-6; Neutrophil S100 protein; p6; Protein S100 A12; protein S100-A12; S100 calcium binding protein A12; S100 calcium-binding protein A12; S100 calcium-binding protein A12 (calgranulin C); S100A12;
Gene ID (Entrez) 6283
Gene Symbol S100A12
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target S100A12
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.