Learn More
Raybiotech Inc Human EN-RAGE (S100 A12) ELISA Kit
Supplier:  Raybiotech Inc ELHENRGE1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.82 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human EN-RAGE. Other species not yet determined.
Catalog No. 50-194-9220