Learn More
BIOHIPPO Human ENA78 Microsample
SDP
Catalog No. 5000314536
Encompass
Human ENA78 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ENA78 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ENA78 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ENA78 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ENA78 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14536 Supplier BIOHIPPO Supplier No. BHE15212275
Human ENA78 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ENA78 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ENA78 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ENA78 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ENA78 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color