Learn More
Biomatik Corporation Human Endomorphin 2 (EM2) ELISA Kit
Supplier:  Biomatik Corporation EKU08695
Encompass_Preferred
Human Endomorphin 2 (EM2) ELISA Kit for detecting EM2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.70-300pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: EP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7714