Learn More
Biomatik Corporation Human Endonuclease Domain Containing Protein 1 (ENDOD1) ELISA Kit
Supplier:  Biomatik Corporation EKU09383
Encompass_Preferred
Human Endonuclease Domain Containing Protein 1 (ENDOD1) ELISA Kit for detecting ENDOD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94919. Gene ID: 23052. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8528