Learn More
Biomatik Corporation Human Endonuclease G, Mitochondrial (ENDOG) ELISA Kit
Supplier:  Biomatik Corporation EKU03875
Encompass_Preferred
Human Endonuclease G, Mitochondrial (ENDOG) ELISA Kit for detecting ENDOG in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14249. Gene ID: 2021. Target Synonyms: ENDO-G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6454