Learn More
Biomatik Corporation Human Endoplasmic Reticulum Lipid Raft Associated Protein 2 (ERLIN2) ELISA Kit
Supplier:  Biomatik Corporation EKU03958
Encompass_Preferred
Human Endoplasmic Reticulum Lipid Raft Associated Protein 2 (ERLIN2) ELISA Kit for detecting ERLIN2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94905. Gene ID: 11160. Target Synonyms: Erlin-2; SPFH2; NET32; C8orf2; ER Lipid Raft Associated Protein 2; Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6851