Learn More
Biomatik Corporation Human Endoplasmic Reticulum To Nucleus Signalling 1 (ERN1) ELISA Kit
Supplier:  Biomatik Corporation EKU03877
Encompass_Preferred
Human Endoplasmic Reticulum To Nucleus Signalling 1 (ERN1) ELISA Kit for detecting ERN1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75460. Gene ID: 2081. Target Synonyms: IRE1; IRE1P; Serine/Threonine-Protein Kinase/Endoribonuclease IRE1; Inositol-requiring protein 1; Ire1-alpha; Serine/threonine-protein kinase; Endoribonuclease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4844