Learn More
Biomatik Corporation Human Endothelial Cell Adhesion Molecule (ESAM) ELISA Kit
Supplier:  Biomatik Corporation EKU03882
Encompass_Preferred
Human Endothelial Cell Adhesion Molecule (ESAM) ELISA Kit for detecting ESAM in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96AP7. Gene ID: 90952. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6494