Learn More
Biomatik Corporation Human Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit
Supplier:  Biomatik Corporation EKL57077
Encompass_Preferred
Human Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit for detecting ESM1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: Q9NQ30. Gene ID: 11082. Target Synonyms: Endocan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9161