Learn More
Biomatik Corporation Human Endothelin 1 (EDN1) CLIA Kit
Supplier:  Biomatik Corporation EKU09462
Encompass_Preferred
Human Endothelin 1 (EDN1) CLIA Kit for detecting EDN1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.9-1,000pg/ml. Sensitivity: 1.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P05305. Gene ID: 1906. Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8514