Learn More
Raybiotech Inc Human Endothelin-1 ELISA Kit
Supplier:  Raybiotech Inc ELHEDN11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Endothelin-1 (EDN1)
Catalog No. 50-194-9210