Learn More
Biomatik Corporation Human Endothelin 3 (EDN3) ELISA Kit
Catalog No. 502417473
Encompass_Preferred
Human Endothelin 3 (EDN3) ELISA Kit for detecting EDN3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.46pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P14138. Gene ID: 1908. Target Synonyms: ET3; PPET3; Preproendothelin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7473 Supplier Biomatik Corporation Supplier No. EKU12484
Human Endothelin 3 (EDN3) ELISA Kit for detecting EDN3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.46pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P14138. Gene ID: 1908. Target Synonyms: ET3; PPET3; Preproendothelin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.